5-Bromo-4-fluoro-2,3-dihydro-1H-inden-1-one 96%

Supplier: Ambeed
Warning

A394427-100MG A394427-1G A394427-250MG
77155-530EA 12 USD
77155-530 77155-532 77155-534
5-Bromo-4-fluoro-2,3-dihydro-1H-inden-1-one 96%
5-Bromo-4-fluoro-2,3-dihydro-1H-inden-1-one

Precaución: for Research Use Only

Formula: C₉H₆BrFO
MDL Number: MFCD18208171
CAS Number: 127425-74-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR