4-Fluoro-1-(triisopropylsilyl)-1H-pyrrolo[2,3-b]pyridine 97%

Supplier: Ambeed
Warning

A394058-100MG A394058-1G A394058-250MG
77155-518EA 29.87 USD
77155-518 77155-520 77155-522
4-Fluoro-1-(triisopropylsilyl)-1H-pyrrolo[2,3-b]pyridine 97%
4-Fluoro-1-(triisopropylsilyl)-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₁₆H₂₅FN₂Si
MW: 292.46 g/mol
MDL Number: MFCD11616419
CAS Number: 640735-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR