(2-Cyano-3-fluorophenyl)boronic acid 95+%

Supplier: Ambeed
Warning

A393578-1G A393578-250MG A393578-5G
77155-512EA 207 USD
77155-512 77155-514 77155-516
(2-Cyano-3-fluorophenyl)boronic acid 95+%
(2-Cyano-3-fluorophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₅BFNO₂
MDL Number: MFCD10696669
CAS Number: 1218993-03-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR