Diethylphosphatoethyltriethoxysilane 95%

Supplier: Ambeed
Warning

A393528-100G A393528-25G A393528-5G
77155-506EA 103 USD
77155-506 77155-508 77155-510
Diethylphosphatoethyltriethoxysilane 95%
Diethylphosphatoethyltriethoxysilane

Precaución: for Research Use Only

Formula: C₁₂H₂₉O₆PSi
MDL Number: MFCD00054814
CAS Number: 757-44-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR