Potassium (4-acetamidophenyl)trifluoroboranuide 95+%

Supplier: Ambeed
Warning

A393087-250MG A393087-5G
77155-502EA 11.33 USD
77155-502 77155-504
Potassium (4-acetamidophenyl)trifluoroboranuide 95+%
Potassium (4-acetamidophenyl)trifluoroboranuide

Precaución: for Research Use Only

Formula: C₈H₈BF₃KNO
MW: 241.06 g/mol
MDL Number: MFCD09993541
CAS Number: 1427323-42-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR