4,6-Dibromo-2,3-dichloroaniline 98+%

Supplier: Ambeed
Warning

A391861-1G A391861-25G A391861-5G
77155-486EA 11.33 USD
77155-486 77155-488 77155-490
4,6-Dibromo-2,3-dichloroaniline 98+%
4,6-Dibromo-2,3-dichloroaniline

Precaución: for Research Use Only

Formula: C₆H₃Br₂Cl₂N
MW: 319.81 g/mol
MDL Number: MFCD00052815
CAS Number: 113571-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR