(2-Fluoro-6-methoxypyridin-3-yl)boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-6-methoxypyridine-3-boronic acid

A391183-100MG A391183-10G A391183-1G A391183-250MG A391183-5G
77155-484EA 22 USD
77155-484 77901-078 77901-921 77901-349 77901-376
(2-Fluoro-6-methoxypyridin-3-yl)boronic acid 98%
(2-Fluoro-6-methoxypyridin-3-yl)boronic acid

Precaución: for Research Use Only

Formula: C₆H₇BFNO₃
MDL Number: MFCD16611244
CAS Number: 1402238-30-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR