6-Methylpyridazin-3(2H)-one Hydrobromide 95%

Supplier: Ambeed
Warning

A390940-100G A390940-10G A390940-25G A390940-5G
77155-476EA 146.26 USD
77155-476 77155-478 77155-480 77155-482
6-Methylpyridazin-3(2H)-one Hydrobromide 95%
6-Methylpyridazin-3(2H)-one Hydrobromide

Precaución: for Research Use Only

Formula: C₅H₇BrN₂O
MW: 191.03 g/mol
MDL Number: MFCD16620340
CAS Number: 94248-99-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR