2-Fluoro-3-(trifluoromethoxy)benzoic acid 98%

Supplier: Ambeed
Warning

A390619-1G A390619-250MG A390619-25G
77155-460EA 44.29 USD
77155-460 77155-462 77155-464
2-Fluoro-3-(trifluoromethoxy)benzoic acid 98%
2-Fluoro-3-(trifluoromethoxy)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₄F₄O₃
MW: 224.109 g/mol
MDL Number: MFCD12026477
CAS Number: 1159512-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR