2-(3-Bromo-5-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Bromo-5-methoxyphenylboronic acid pinacol ester
A390378-100MG
A390378-1G
A390378-250MG
A390378-5G
77155-452EA
16.48
USD
77155-452
77155-454
77155-456
77155-458
2-(3-Bromo-5-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%
2-(3-Bromo-5-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Precaución: for Research Use Only
Formula:
C₁₃H₁₈BBrO₃ MW: 312.995 g/mol |
MDL Number:
MFCD12405339 CAS Number: 401797-04-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...