7-(4-Bromobutoxy)quinolin-2(1H)-one 95%

Supplier: Ambeed
Warning

A389894-1G A389894-250MG A389894-25G A389894-5G
77155-444EA 19.57 USD
77155-444 77155-446 77155-448 77155-450
7-(4-Bromobutoxy)quinolin-2(1H)-one 95%
7-(4-Bromobutoxy)quinolin-2(1H)-one

Precaución: for Research Use Only

Formula: C₁₃H₁₄BrNO₂
MW: 296.16 g/mol
MDL Number: MFCD09753615
CAS Number: 203395-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR