[4-(Phenyl-BOC-aminomethyl)phenyl] boronic acid pinacol ester 95+%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl phenyl(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)carbamate

A388596-1G A388596-250MG A388596-5G
77155-426EA 143 USD
77155-426 77155-428 77155-430
[4-(Phenyl-BOC-aminomethyl)phenyl] boronic acid pinacol ester 95+%
[4-(Phenyl-BOC-aminomethyl)phenyl] boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₂₄H₃₂BNO₄
MW: 409.326 g/mol
MDL Number: MFCD09953500
CAS Number: 1073371-71-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR