Methyl benzothiophene-3-carboxylate 98%

Supplier: Ambeed
Warning

A386169-100MG A386169-1G A386169-250MG A386169-5G
77155-404EA 33 USD
77155-404 77155-406 77155-408 77155-410
Methyl benzothiophene-3-carboxylate 98%
Methyl benzothiophene-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₈O₂S
MDL Number: MFCD06203855
CAS Number: 22913-25-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR