6-Hydrazinoquinoline dihydrochloride 95%

Supplier: Ambeed
Warning

A385282-1G A385282-250MG A385282-5G
77155-400EA 25 USD
77155-400 77155-402 77900-972
6-Hydrazinoquinoline dihydrochloride 95%
6-Hydrazinoquinoline dihydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₁Cl₂N₃
MW: 232.11 g/mol
MDL Number: MFCD22123347
CAS Number: 103755-52-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR