trans-1-Boc-3-fluoro-4-hydroxypiperidine 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl trans-3-fluoro-4-hydroxypiperidine-1-carboxylate

A384757-100MG A384757-1G A384757-250MG
77155-394EA 57.68 USD
77155-394 77155-396 77155-398
trans-1-Boc-3-fluoro-4-hydroxypiperidine 97%
trans-1-Boc-3-fluoro-4-hydroxypiperidine

Precaución: for Research Use Only

Formula: C₁₀H₁₈FNO₃
MW: 219.26 g/mol
MDL Number: MFCD18791210
CAS Number: 955029-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR