2-(3-Nitrophenyl)-1,3-dioxolane 95%

Supplier: Ambeed
Warning

A384652-100MG A384652-1G A384652-250MG A384652-5G
77155-386EA 6 USD
77155-386 77155-388 77155-390 77155-392
2-(3-Nitrophenyl)-1,3-dioxolane 95%
2-(3-Nitrophenyl)-1,3-dioxolane

Precaución: for Research Use Only

Formula: C₉H₉NO₄
MDL Number: MFCD00020899
CAS Number: 6952-67-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR