Diethylene Glycol Mono(2-propyn-1-yl) ether 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-[2-(2-Propynyloxy)ethoxy]ethanol, 2-[2-(2-Propyn-1-yloxy)ethoxy]ethanol, Propargyl-PEG2-OH
A383690-100MG
A383690-1G
A383690-250MG
A383690-5G
A383690-25G
77155-356EA
6
USD
77155-356
77155-358
77155-360
77155-362
77686-068
Diethylene Glycol Mono(2-propyn-1-yl) ether 97%
Diethylene Glycol Mono(2-propyn-1-yl) ether
Precaución: for Research Use Only
Formula:
C₇H₁₂O₃ MW: 144.17 g/mol |
MDL Number:
MFCD00053353 CAS Number: 7218-43-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...