Sodium pyruvate 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt
A383017-1000G
A383017-100G
A383017-25G
A383017-500G
77155-346EA
51
USD
77155-346
77155-348
77155-350
77155-352
Sodium pyruvate 99%
Sodium pyruvate
Precaución: for Research Use Only
Formula:
CH₃COCOONa MW: 110.04 g/mol |
MDL Number:
MFCD00002586 CAS Number: 113-24-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...