Tin(II) acetate 95%

Supplier: Ambeed
Danger

Synonyms: Tin(II) Acetate

A382331-100G A382331-25G A382331-5G
77155-330EA 43 USD
77155-330 77155-332 77155-334
Tin(II) acetate 95%
Tin(II) acetate

Precaución: for Research Use Only

Formula: C₄H₆O₄Sn
MW: 236.8 g/mol
MDL Number: MFCD00054408
CAS Number: 638-39-1
UN: 3146
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR