CCG-1423 98%
Supplier: Ambeed
|
Danger
|
A381687-10MG
A381687-1MG
A381687-50MG
A381687-5MG
77155-322EA
19
USD
77155-322
77155-324
77155-326
77155-328
CCG-1423 98%
CCG-1423
Precaución: for Research Use Only
Formula:
C18H13ClF6N2O3 |
MDL Number:
MFCD01566719 CAS Number: 285986-88-1 UN: 1325 ADR: 4.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...