Ethyl-3-benzyl-3-azabicyclo[3.1.0]hexane-1-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-benzyl-3-azabicyclo[3.1.0]hexane-1-carboxylate

A379762-100MG A379762-250MG A379762-5G
77155-298EA 39.14 USD
77155-298 77155-300 77155-302
Ethyl-3-benzyl-3-azabicyclo[3.1.0]hexane-1-carboxylate 95%
Ethyl-3-benzyl-3-azabicyclo[3.1.0]hexane-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₅H₁₉NO₂
MW: 245.32 g/mol
MDL Number: MFCD22566243
CAS Number: 63618-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR