Ethyl-6-bromo-4-hydroxyquinoline-3-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 6-bromo-4-hydroxyquinoline-3-carboxylate

A379650-100G A379650-10G A379650-1G A379650-250MG A379650-25G A379650-5G
77155-286EA 520.15 USD
77155-286 77155-288 77155-290 77155-292 77155-294 77155-296
Ethyl-6-bromo-4-hydroxyquinoline-3-carboxylate 95%
Ethyl 6-bromo-4-hydroxyquinoline-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₀BrNO₃
MW: 296.1200 g/mol
MDL Number: MFCD00173362
CAS Number: 122794-99-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR