(2'-Fluoro-[1,1'-biphenyl]-4-yl)boronic acid 95%

Supplier: Ambeed
Warning

A378370-100MG A378370-1G A378370-250MG
77155-270EA 320.33 USD
77155-270 77155-272 77155-274
(2'-Fluoro-[1,1'-biphenyl]-4-yl)boronic acid 95%
(2'-Fluoro-[1,1'-biphenyl]-4-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₀BFO₂
MW: 216.02 g/mol
MDL Number: MFCD03095260
CAS Number: 1383531-49-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR