1-Bromo-4-chloro-2-fluorobenzene 98%

Supplier: Ambeed
Warning

A376375-1000G A376375-100G A376375-10G A376375-25G A376375-500G A376375-5G
77155-250EA 337.01 USD
77155-250 77155-252 77155-254 77155-256 77155-258 77155-260
1-Bromo-4-chloro-2-fluorobenzene 98%
1-Bromo-4-chloro-2-fluorobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrClF
MDL Number: MFCD00079708
CAS Number: 1996-29-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR