Methyl piperazine-2-carboxylate, N1-CBZ protected 97%

Supplier: Ambeed
Warning

Synonyms: 1-Benzyl 2-methyl piperazine-1,2-dicarboxylate, Piperazine-2-carboxylic acid methyl ester, N1-CBZ protected

A374423-1MG A374423-250MG A374423-1G
77155-242EA 5.15 USD
77155-242 77155-244 77686-045
Methyl piperazine-2-carboxylate, N1-CBZ protected 97%
Methyl piperazine-2-carboxylate, N1-CBZ protected

Precaución: for Research Use Only

Formula: C₁₄H₁₈N₂O₄
MW: 278.304 g/mol
MDL Number: MFCD04115344
CAS Number: 126937-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR