AR-C155858 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-6-((3,5-Dimethyl-1H-pyrazol-4-yl)methyl)-5-(4-hydroxyisoxazolidine-2-carbonyl)-1-isobutyl-3-methylthieno[2,3-d]pyrimidine-2,4(1H,3H)-dione, 6-[(3,5-Dimethyl-1H-pyrazol-4-yl)methyl]-5-[[(4S)-4-hydroxy-2-isoxazolidinyl]carbonyl]-3-methyl-1-(2-methylpropyl)thieno[2,3-d]pyrimidine-2,4(1H,3H)-dione
A373794-10MG
A373794-1MG
77155-238EA
497.49
USD
77155-238
77155-240
AR-C155858 98%
AR-C155858
Precaución: for Research Use Only
Formula:
C₂₁H₂₇N₅O₅S MW: 461.53 g/mol |
MDL Number:
MFCD22124476 CAS Number: 496791-37-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...