BrettPhos palladacycle 3rd generation 97%

Supplier: Ambeed
Warning

Synonyms: [(2-Di-cyclohexylphosphino-3,6-dimethoxy-2′,4′,6′- triisopropyl-1,1′-biphenyl)-2-(2′-amino-1,1′ -biphenyl)]palladium(II) methanesulphonate, BrettPhos Pd G3

A371066-100MG A371066-10G A371066-1G A371066-5G A371066-250MG A371066-25G
77155-220EA 11.33 USD
77155-220 77155-222 77155-224 77155-226 77613-102 77613-104
BrettPhos palladacycle 3rd generation 97%
BrettPhos palladacycle 3rd generation

Precaución: for Research Use Only

Formula: C₄₈H₆₆NO₅PPdS
MW: 906.50 g/mol
MDL Number: MFCD22572666
CAS Number: 1470372-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR