8-Fluoroquinoline-3-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 3-Carboxy-8-fluoroquinoline

A370764-100MG A370764-1G A370764-250MG A370764-5G
77155-212EA 13.39 USD
77155-212 77155-214 77155-216 77155-218
8-Fluoroquinoline-3-carboxylic acid 95%
8-Fluoroquinoline-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₆FNO₂
MW: 191.159 g/mol
MDL Number: MFCD11108738
CAS Number: 71082-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR