1-Hexyl-3-methylimidazolium tetrafluoroborate 97%

Supplier: Ambeed
Warning

A370499-100G A370499-25G
77155-208EA 68 USD
77155-208 77155-210
1-Hexyl-3-methylimidazolium tetrafluoroborate 97%
1-Hexyl-3-methylimidazolium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₁₀H₁₉BF₄N₂
MDL Number: MFCD03095459
CAS Number: 244193-50-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR