Tetrabutylammonium triiodide 96%

Supplier: Ambeed
Warning

Synonyms: Tetrabutylammonium Triiodide

A369111-1G A369111-5G A369111-25G
77155-204EA 35 USD
77155-204 77686-039 77900-605
Tetrabutylammonium triiodide 96%
Tetrabutylammonium triiodide

Precaución: for Research Use Only

Formula: C₁₆H₃₆I₃N
MW: 623.18 g/mol
MDL Number: MFCD00059123
CAS Number: 13311-45-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR