tert-Butyl 7-oxo-5-oxa-2-azaspiro[3.4]octane-2-carboxylate ≥97%

Supplier: Ambeed
Danger

A368303-100MG A368303-1G A368303-250MG A368303-5G A368303-25G
77155-196EA 11 USD
77155-196 77155-198 77155-200 77155-202 77572-100
tert-Butyl 7-oxo-5-oxa-2-azaspiro[3.4]octane-2-carboxylate ≥97%
tert-Butyl 7-oxo-5-oxa-2-azaspiro[3.4]octane-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₇NO₄
MDL Number: MFCD23105989
CAS Number: 1408075-90-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR