(Z)-3-(2-Phenylhydrazono)cyclohexanone 95%

Supplier: Ambeed
Danger

A367615-250MG A367615-100MG
77155-194EA 81 USD
77155-194 77900-856
(Z)-3-(2-Phenylhydrazono)cyclohexanone 95%
(Z)-3-(2-Phenylhydrazono)cyclohexanone

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂O
MDL Number: MFCD11559082
UN: 2811
ADR: 6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR