1,1,3,3-Tetramethyl-2-(perfluorophenyl)isouronium hexafluorophosphate(V) 98%

Supplier: Ambeed
Warning

A364516-25G A364516-5G
77155-162EA 281.19 USD
77155-162 77155-164
1,1,3,3-Tetramethyl-2-(perfluorophenyl)isouronium hexafluorophosphate(V) 98%
1,1,3,3-Tetramethyl-2-(perfluorophenyl)isouronium hexafluorophosphate(V)

Precaución: for Research Use Only

Formula: C₁₁H₁₂F₁₁N₂OP
MW: 428.18 g/mol
MDL Number: MFCD04973269
CAS Number: 206190-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR