6-Bromo-7-fluoroisoquinoline 95%

Supplier: Ambeed
Warning

A362781-1G A362781-250MG
77155-144EA 48 USD
77155-144 77892-744
6-Bromo-7-fluoroisoquinoline 95%
6-Bromo-7-fluoroisoquinoline

Precaución: for Research Use Only

Formula: C₉H₅BrFN
MDL Number: MFCD28166295
CAS Number: 1258833-80-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR