(2S,3R)-3-(2,5-Difluorophenyl)-3-hydroxy-2-methyl-4-(1H-1,2,4-triazol-1-yl)butanenitrile 95%

Supplier: Ambeed
Warning

A362605-1G A362605-5G
77155-140EA 43.26 USD
77155-140 77155-142
(2S,3R)-3-(2,5-Difluorophenyl)-3-hydroxy-2-methyl-4-(1H-1,2,4-triazol-1-yl)butanenitrile 95%
(2S,3R)-3-(2,5-Difluorophenyl)-3-hydroxy-2-methyl-4-(1H-1,2,4-triazol-1-yl)butanenitrile

Precaución: for Research Use Only

Formula: C₁₃H₁₂F₂N₄O
MW: 278.25 g/mol
MDL Number: MFCD28411477
CAS Number: 241479-74-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR