2,6-Dimethylbenzoyl chloride 95%

Supplier: Ambeed
Danger

A362593-100MG A362593-250MG
77155-136EA 20.6 USD
77155-136 77155-138
2,6-Dimethylbenzoyl chloride 95%
2,6-Dimethylbenzoyl chloride

Precaución: for Research Use Only

Formula: C₉H₉ClO
MW: 168.62 g/mol
MDL Number: MFCD00052854
CAS Number: 21900-37-8
UN: 3265
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR