1,2-Bis((R)-tert-butyl(methyl)phosphino)benzene 98%
Supplier: Ambeed
|
Warning
|
A362120-100MG
A362120-250MG
A362120-50MG
A362120-1G
77155-130EA
152.44
USD
77155-130
77155-132
77155-134
77568-734
1,2-Bis((R)-tert-butyl(methyl)phosphino)benzene 98%
1,2-Bis((R)-tert-butyl(methyl)phosphino)benzene
Precaución: For research use only.
Formula:
C₁₆H₂₈P₂ MW: 282.34 g/mol |
MDL Number:
MFCD18837015 CAS Number: 919778-41-9 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...