(6R,7S,12AS)-1,2,11,12-tetramethoxy-6,7-dimethyl-5,6,7,8-tetrahydrodibenzo[a,c][8]annulene-3,10-diol 95+%

Supplier: Ambeed
Warning

Synonyms: Gomisin J

A361884-1MG A361884-5MG
77155-126EA 49.44 USD
77155-126 77155-128
(6R,7S,12AS)-1,2,11,12-tetramethoxy-6,7-dimethyl-5,6,7,8-tetrahydrodibenzo[a,c][8]annulene-3,10-diol 95+%
(6R,7S,12AS)-1,2,11,12-tetramethoxy-6,7-dimethyl-5,6,7,8-tetrahydrodibenzo[a,c][8]annulene-3,10-diol

Precaución: for Research Use Only

Formula: C₂₂H₂₈O₆
MW: 388.45 g/mol
MDL Number: MFCD29048562
CAS Number: 66280-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR