4,6-Dimethoxypyridin-3-amine 97%

Supplier: Ambeed
Warning

A361812-100MG A361812-250MG
77155-122EA 45.32 USD
77155-122 77155-124
4,6-Dimethoxypyridin-3-amine 97%
4,6-Dimethoxypyridin-3-amine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O₂
MW: 154.16 g/mol
MDL Number: MFCD09027594
CAS Number: 89943-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR