trans-4-n-Propyl-4'-(3,4,5-trifluorophenyl)bicyclohexyl 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
trans,trans-4'-Propyl-4-(3,4,5-trifluorophenyl)bicyclohexyl
A360723-1G
A360723-5G
77155-108EA
8.24
USD
77155-108
77155-110
trans-4-n-Propyl-4'-(3,4,5-trifluorophenyl)bicyclohexyl 97%
trans-4-n-Propyl-4'-(3,4,5-trifluorophenyl)bicyclohexyl
Precaución: for Research Use Only
Formula:
C₂₁H₂₉F₃ MW: 338.46 g/mol |
MDL Number:
MFCD09838998 CAS Number: 131819-23-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...