2-Chloro-5-(trichloromethyl)pyridine 95%

Supplier: Ambeed
Warning

A360261-1G A360261-25G
77155-094EA 5.15 USD
77155-094 77155-096
2-Chloro-5-(trichloromethyl)pyridine 95%
2-Chloro-5-(trichloromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₃Cl₄N
MDL Number: MFCD00160145
CAS Number: 69045-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR