1-Iodooctane 98% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilized with Copper chip

A359564-500G A359564-5G A359564-25G A359564-100G
77155-090EA 172 USD
77155-090 77155-092 77572-062 77901-500
1-Iodooctane 98% stabilized
1-Iodooctane

Precaución: for Research Use Only

Formula: C₈H₁₇I
MDL Number: MFCD00001105
CAS Number: 629-27-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR