Diphenylvinylsulfonium triflate 98%

Supplier: Ambeed
Warning

Synonyms: Diphenyl(vinyl)sulphonium trifluoromethanesulphonate

A359353-1G A359353-250MG A359353-5G A359353-25G
77155-078EA 27 USD
77155-078 77155-080 77551-022 77686-017
Diphenylvinylsulfonium triflate 98%
Diphenylvinylsulfonium triflate

Precaución: for Research Use Only

Formula: C₁₅H₁₃F₃O₃S₂
MW: 362.39 g/mol
MDL Number: MFCD22123284
CAS Number: 247129-88-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR