1-Benzylindole-5-boronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 1-Benzyl-5-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-1H-indole

A358662-1G A358662-250MG A358662-5G
77155-066EA 147 USD
77155-066 77155-068 77155-070
1-Benzylindole-5-boronic acid pinacol ester 98%
1-Benzylindole-5-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₂₁H₂₄BNO₂
MW: 333.24 g/mol
MDL Number: MFCD11894320
CAS Number: 1206163-56-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR