(S)-(-)-1-Amino-2-(methoxymethyl)pyrrolidine 98%

Supplier: Ambeed
Warning

A358384-100MG A358384-1G A358384-250MG A358384-5G
77155-058EA 11 USD
77155-058 77155-060 77155-062 77155-064
(S)-(-)-1-Amino-2-(methoxymethyl)pyrrolidine 98%
(S)-(-)-1-Amino-2-(methoxymethyl)pyrrolidine

Precaución: for Research Use Only

Formula: C₆H₁₄N₂O
MDL Number: MFCD00064485
CAS Number: 59983-39-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR