3,5-Dibromo-2-methoxypyridine 98%

Supplier: Ambeed
Warning

A358350-1G A358350-250MG A358350-25G A358350-5G
77155-050EA 6.18 USD
77155-050 77155-052 77155-054 77155-056
3,5-Dibromo-2-methoxypyridine 98%
3,5-Dibromo-2-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅Br₂NO
MW: 266.9200 g/mol
MDL Number: MFCD05663709
CAS Number: 13472-60-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR