Ethyl-5-fluoro-2-methoxybenzoylformate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 5-fluoro-2-methoxybenzoylformate, Ethyl 2-(5-fluoro-2-methoxyphenyl)-2-oxoacetate

A356665-100MG A356665-1G A356665-250MG
77155-036EA 48.41 USD
77155-036 77155-038 77155-040
Ethyl-5-fluoro-2-methoxybenzoylformate 95%
Ethyl-5-fluoro-2-methoxybenzoylformate

Precaución: for Research Use Only

Formula: C₁₁H₁₁FO₄
MW: 226.21 g/mol
MDL Number: MFCD09801422
CAS Number: 873548-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR