(3-Bromo-1-propyn-1-yl)cyclopropane 95%

Supplier: Ambeed
Warning

A353668-100MG A353668-250MG A353668-1G A353668-5G
77155-016EA 36.05 USD
77155-016 77155-018 77686-006 77686-007
(3-Bromo-1-propyn-1-yl)cyclopropane 95%
(3-Bromo-1-propyn-1-yl)cyclopropane

Precaución: for Research Use Only

Formula: C₆H₇Br
MW: 159.02 g/mol
MDL Number: MFCD20623445
CAS Number: 852526-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR