4-Fluoro-[1,1'-biphenyl]-3-amine 95%

Supplier: Ambeed
Warning

A353020-100MG A353020-250MG
77155-010EA 51.5 USD
77155-010 77155-012
4-Fluoro-[1,1'-biphenyl]-3-amine 95%
4-Fluoro-[1,1'-biphenyl]-3-amine

Precaución: for Research Use Only

Formula: C₁₂H₁₀FN
MW: 187.21 g/mol
MDL Number: MFCD14701577
CAS Number: 478403-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR