Methyl 3-aminopyridazine-4-carboxylate 97%

Supplier: Ambeed
Warning

A352422-100MG A352422-1G
77155-006EA 157 USD
77155-006 77155-008
Methyl 3-aminopyridazine-4-carboxylate 97%
Methyl 3-aminopyridazine-4-carboxylate

Precaución: for Research Use Only

Formula: C₆H₇N₃O₂
MDL Number: MFCD18071012
CAS Number: 1256633-18-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR